Merck
  • Synthesis, structural, spectral, thermal and antimicrobial studies of palladium(II), platinum(II), ruthenium(III) and iridium(III) complexes derived from N,N,N,N-tetradentate macrocyclic ligand.
Synthesis, structural, spectral, thermal and antimicrobial studies of palladium(II), platinum(II), ruthenium(III) and iridium(III) complexes derived from N,N,N,N-tetradentate macrocyclic ligand.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-02-22)
Soni Rani, Sumit Kumar, Sulekh Chandra
ABSTRACT
Palladium(II), platinum(II), ruthenium(III) and iridium(III) complexes of general stoichiometry [PdL]Cl(2), [PtL]Cl(2), [Ru(L)Cl(2)]Cl and [Ir(L)Cl(2)]Cl are synthesized with a tetradentate macrocyclic ligand, derived from 2,6-diaminopyridine with 3-ethyl 2,4-pentanedione. Ligand was characterized on the basis of elemental analyses, IR, mass, and (1)H NMR and (13)C NMR spectral studies. All the complexes were characterized by elemental analyses, molar conductance measurements, magnetic susceptibility measurements, IR, mass, electronic spectral techniques and thermal studies. The value of magnetic moments indicates that all the complexes are diamagnetic except Ru(III) complex which shows magnetic moments corresponding its one unpaired electron. The macrocyclic ligand and all its metal complexes were also evaluated in vitro against some plant pathogenic fungi and bacteria to assess their biocidal properties.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%