- Synthesis of ceramides using N-hydroxysuccinimide esters.
Synthesis of ceramides using N-hydroxysuccinimide esters.
Journal of lipid research (1972-11-01)
D E Ong, R N Brady
PMID4641422
ABSTRACT
Fatty acyl esters of N-hydroxysuccinimide have been used to directly N-acylate sphingenine or sphinganine, forming the corresponding ceramides. The reaction proceeds in excellent yield (84-96%) from small amounts of starting material (10-20 mg). The product ceramides are pure after one recrystallization.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto