Merck
  • Homoleptic aminophenolates of Zn, Mg and Ca. Synthesis, structure, DFT studies and polymerization activity in ROP of lactides.
Homoleptic aminophenolates of Zn, Mg and Ca. Synthesis, structure, DFT studies and polymerization activity in ROP of lactides.
Dalton transactions (Cambridge, England : 2003) (2013-12-07)
Jakub Wojtaszak, Krzysztof Mierzwicki, Sławomir Szafert, Nurbey Gulia, Jolanta Ejfler
ABSTRACT
The reaction of MgBu2, ZnEt2 or Ca(O(i)Pr)2 with 2 eq. of three-coordinating N-[methyl(2-hydroxy-3,5-dimethylphenyl)]-N-methyl-N-methyl-1,3-oxolaneamine (mpoa-H) or N-[methyl(2-hydroxy-3,5-di-tert-butylphenyl)]-N-methyl-N-methyl-1,3-oxolaneamine (tbpoa-H) gave neutral, monomeric [Mg(mpoa)2], [Zn(mpoa)2], [Zn(tbpoa)2], and [Ca(tbpoa)2] as white powders in 58-90% yields. The resulting aminophenolates were characterized in solution by NMR showing, in the case of [Zn(tbpoa)2], interesting dynamics. [Zn(tbpoa)2] and [Ca(tbpoa)2] were characterized by X-ray crystallography to show the Zn atom to be pseudo-octahedrally coordinated and the Ca atom in six-coordination mode. The new homoleptic complexes were tested in the polymerization of lactide with an external alcohol to reveal stable behaviour (during the polymerization process) only in the case of [Zn(tbpoa)2]. The high activity of the catalyst was correlated with a ligand flexibility that was further supported by theoretical studies.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-Methylaminomethyl-1,3-dioxolane, 98%