Merck
Lactoferrin administration into the nostril alleviates murine allergic rhinitis and its mechanisms.
Scandinavian journal of immunology (2013-10-12)
S B Wang, Y Q Deng, J Ren, B K Xiao, Z Chen, Z Z Tao
ABSTRACT
Lactoferrin (LF) can downregulate allergic airway inflammation in asthma. However, the in vivo effect of exogenous LF on allergic rhinitis (AR), a disease attributed to airway inflammation, has yet to be determined. We investigated the effect of intranasal administration recombinant human (rh) LF and its underlying mechanisms on AR in BALB/c mice. Multiple parameters of allergic responses were evaluated to determine the effect of rhLF. We found that the number of eosinophils and goblet cells, as well as mRNA and protein expression of type 2 helper T (Th2), Th17 and regulatory T (Treg) cells in the nasal cavity, was significantly upregulated in AR mice compared with the controls, Conversely, administration of rhLF prior to or after intranasal ovalbumin challenge markedly downregulated these same parameters. Th1-specific mRNA and protein expression in the nasal cavity of the controls was not different from that in AR mice, but expression significantly increased with rhLF treatment. The mRNA and protein expression of endogenous LF in the nasal cavity was significantly downregulated in AR mice compared with the controls. However, after rhLF treatment, endogenous LF mRNA and protein expression was significantly upregulated. Exogenous rhLF inhibited allergic inflammation in AR mice, most likely by promoting the endogenous LF expression and skewing T cells to a Th1, but not a Th2 and Th17 phenotype in the nasal mucosa. Our findings suggest that rhLF treatment may be a novel therapeutic approach for prevention and treatment AR.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Albumina, lyophilized powder, ≥98% (agarose gel electrophoresis)
SKU
Taglio della confezione
Disponibilità
Prezzo
Quantità
Sigma-Aldrich
Albumina, lyophilized powder, ≥98% (agarose gel electrophoresis)
SKU
Taglio della confezione
Disponibilità
Prezzo
Quantità
Sigma-Aldrich
Albumina, powder, 62-88% (agarose gel electrophoresis)
SKU
Taglio della confezione
Disponibilità
Prezzo
Quantità
Sigma-Aldrich
Albumina, lyophilized powder, ≥90% (agarose gel electrophoresis)
SKU
Taglio della confezione
Disponibilità
Prezzo
Quantità
Sigma-Aldrich
Albumina, lyophilized powder
SKU
Taglio della confezione
Disponibilità
Prezzo
Quantità
Supelco
Albumina, For use as a marker in SDS-PAGE
SKU
Taglio della confezione
Disponibilità
Prezzo
Quantità