Merck
  • RIP1 expression is necessary for CD30-mediated cell death induction in anaplastic large-cell lymphoma cells.
RIP1 expression is necessary for CD30-mediated cell death induction in anaplastic large-cell lymphoma cells.
Laboratory investigation; a journal of technical methods and pathology (2013-04-03)
Burkhard Hirsch, Edda von der Wall, Michael Hummel, Horst Dürkop
ABSTRACT
CD30, a member of the tumor necrosis factor receptor (TNFR) superfamily, is consistently expressed by tumor cells of anaplastic large-cell lymphoma (ALCL). CD30 stimulation induces massive caspase-dependent cell death of ALCL cells in case of canonical NFκB inhibition or proteasome inhibition. However, CD30, a TNFR lacking a death domain (DD), is unable to recruit a death inducing complex containing TRADD (TNFR1-associated DD-protein) or FADD (FAS-associated DD-domain protein) together with the receptor-interacting protein 1 (RIP1) and caspase-8. Thus, the mechanism explaining CD30-induced cell death of lymphocytes remains obscure. Here, we demonstrate that blockage of RIP1 by siRNA or pharmacological inhibition of RIP1 by Necrostatin-1 almost completely prevented CD30-induced cell death. In addition, we revealed CD30-induced accumulation of RIP1 at the cytoplasma membrane of NFκB-inhibited ALCL cells by confocal laser scanning microscopy. Finally, primary ALCL cases can be subdivided into two groups based on the presence or absence of RIP1 as revealed by immunohistology. Taken together, our study identified RIP1 as a crucial mediator of CD30-induced cell death that bears features of apoptosis as well as necroptosis. RIP1 expression in ALCL tumor cells might eligible for the therapeutic application of CD30 antibodies in combination with NFκB/proteasome inhibitors that should result in CD30-induced cell death.