Merck
  • Akathisia--rare cause of psychomotor agitation in patients with traumatic brain injury: case report and review of literature.
Akathisia--rare cause of psychomotor agitation in patients with traumatic brain injury: case report and review of literature.
Journal of rehabilitation research and development (2013-02-15)
Janet E Wielenga-Boiten, Gerard M Ribbers
ABSTRACT
Akathisia is reported to be one of the most common and disabling side effects of antipsychotics and other drugs. Akathisia is also a rare cause of psychomotor agitation in patients with traumatic brain injury (TBI). In this clinical note, we describe a case report of akathisia in a 34-year-old woman with TBI; review earlier case studies on akathisia in TBI; and discuss the differential diagnosis and its pathophysiology, treatment, and prognosis.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Supelco
Clonidine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®