Merck
Effect of lysine vasopressin on pentylenetetrazol-induced retrograde amnesia in rats.
Pharmacology, biochemistry, and behavior (1977-07-01)
H B Bookin, W D Pfeifer
PMID561964
ABSTRACT
Lysine vasopressin (1 microgram/rat SC) administered 1 hr prior to either the acquisition trial or 24 hr retention trial facilitated passive avoidance retention. Amnesia was produced when a single 50 mg/kg (IP) injection of pentylenetetrazol was given immediately following the passive avoidance acquisition trial. A single injection of lysine vasopressin (1 microgram/rat SC) administered 1 hr prior to either the acquisition trial or 24 hr retention trial antagonized the amnesia.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
[Lys8]-Vasopressin, ≥98% (HPLC)