Merck
Inhibition of tyrosinase activity by N,N-unsubstituted selenourea derivatives.
Biological & pharmaceutical bulletin (2005-05-03)
Sang Keun Ha, Mamoru Koketsu, Kunho Lee, Sang Yoon Choi, Ji-Ho Park, Hideharu Ishihara, Sun Yeou Kim
ABSTRACT
This study investigated inhibitory effects of N,N-unsubstituted selenourea derivatives on tyrosinase activity. Three types of N,N-unsubstituted selenoureas derivatives exhibited inhibitory effect on dopa (3,4-dihydroxyphenylalanine) oxidase activity of mushroom tyrosinase. Compound D at a concentration of 200 microM exhibited 55.5% of inhibition on dopa oxidase activity of mushroom tyrosinase. This inhibitory effect was higher than that of kojic acid (39.4%), a well known tyrosinase inhibitor. Moreover, the compound D identified as a noncompetitive inhibitor by Lineweaver-Burk plot analysis. In addition, compound D also inhibited the melanin production in melan-a cells.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Selenourea, 98%