Merck
  • [Determination of evodiamine and rutaecarpine of compound Wuzhuyu cataplasm in plasma by SPE-HPLC: application to its pharmacokinetics].
[Determination of evodiamine and rutaecarpine of compound Wuzhuyu cataplasm in plasma by SPE-HPLC: application to its pharmacokinetics].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-03-10)
Wenguang Yin, Manling Li, Shuzhi Liu, Chen Kang, Yanli Wang, Zhongming Zhou, Huijuan Tong
ABSTRACT
To establish a SPE-HPLC method for the determination and pharmacokinetic study of evodiamine and rutacarpine in rat plasma. A Kromasil C18 column (4.6 mm x 250 mm, 5 microm) was used with acetonitrile-water-tetrahydrofuran-acetic acid (51:48:1:0.1) as a mobile phase and at a flow rate of 1 mL x min(-1), and the UV detection was at 225 nm. The column temperature was 35 degrees C. After the analytes were extracted from the plasma of rats by solid phase extraction (SPE), the content of evodiamine and rutaecarpine was measuared by HPLC method using halcinonide as an internal standard solution. After transdermal administration to rats, the pharmacokinetic behavior of evodiamine and rutaecarpine belongs to the one-compartment model. The main pharmacokinetic parameters was as follows: K(a) 0.224 h(-1) and 0.220 h(-1), K(e) 0.114 h(-1) and 0.118 h(-1), C(max) 0.211 mg x L(-1) and 0.272 mg x L(-1), T(peak) 6.132 h and 6.102 h, respectively. The method in this study is simple, rapid and sensitive. It is proved to be suitable for pharmacokinetic study of evodiamine and rutaecarpine.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Rutaecarpine, >98% (HPLC)