- Modulated growth of nanoparticles. Application for sensing nerve gases.
Modulated growth of nanoparticles. Application for sensing nerve gases.
Analytical chemistry (2008-12-04)
Ana Virel, Laura Saa, Valeri Pavlov
PMID19049371
ABSTRACT
Hydrolysis of acetylthiocholine mediated by acetylcholine esterase yields the thiol-bearing compound thiocholine. At trace concentrations, thiocholine modulates the growth of Au-Ag nanoparticles on seeding gold nanoparticles in the presence of ascorbic acid. Inhibition of the enzyme by 1,5-bis(4-allyldimethylammoniumphenyl)pentan-3-one dibromide (BW284c51) or by diethyl p-nitrophenyl phosphate (paraoxon) produces lower yields of thiocholine, promoting the catalytic growth of Au-Ag nanoparticles. Here, we describe the development of a simple and sensitive colorimetric assay for the detection of AChE inhibitors.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto