Merck
  • Direct ESR detection of pentadienyl radicals and peroxyl radicals in lipid peroxidation: mechanistic insight into regioselective oxygenation in lipoxygenases.
Direct ESR detection of pentadienyl radicals and peroxyl radicals in lipid peroxidation: mechanistic insight into regioselective oxygenation in lipoxygenases.
Journal of the American Chemical Society (2005-05-05)
Hironori Kitaguchi, Kei Ohkubo, Seiji Ogo, Shunichi Fukuzumi
ABSTRACT
Well-resolved ESR spectra of free pentadienyl radicals have been observed under photoirradiation of di-tert-butylperoxide (Bu(t)OOBu(t)) and polyunsaturated fatty acids in the absence of O(2), allowing us to determine the hfc values. The hfc values of linoleyl radical indicate that the spin density is the largest at the C-11 position. The linoleyl radical is readily trapped by O(2) to produce the peroxyl radical (11-HPO.) in which O(2) is added mainly at the C-11 position of the pentadienyl radical as indicated by the comparison of the ESR spectra of peroxyl radicals derived from linoleic acid and [11,11-(2)H(2)]linoleic acid. The peroxyl radical (13-HPO.), which is initially formed by the hydrogen abstraction from 13-(S)-hydroperoxy-9(Z),11(E)-octadecadienoic acid (13-HPOD) by Bu(t)O., is found to isomerize to 11-HPO. via removal of O(2) from 13-HPO. and addition of O(2) to linoleyl radical to produce 11-HPO. . This finding supports an idea of O(2) entering via a specific protein channel, which determines the stereo- and regiochemistry of the biradical combination between O(2) and linoleyl radical in lipoxygenases.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Luperox® DI, tert-Butyl peroxide, 98%