Merck
  • Development of a fully automated on-line solid phase extraction and high-performance liquid chromatography with diode array detection method for the pharmacokinetic evaluation of bavachinin: a study on absolute bioavailability and dose proportionality.
Development of a fully automated on-line solid phase extraction and high-performance liquid chromatography with diode array detection method for the pharmacokinetic evaluation of bavachinin: a study on absolute bioavailability and dose proportionality.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-03-27)
Lei Liu, Kang-Ning Liu, Ya-Bin Wen, Han-Wen Zhang, Ya-Xin Lu, Zheng Yin
ABSTRACT
A fully automated on-line solid-phase extraction (SPE) and high-performance liquid chromatography (HPLC) with diode array detection (DAD) method was developed for determination of bavachinin in mouse plasma. Analytical process was performed on two reversed-phase columns (SPE cartridge and analytical column) connected via a Valco 6-port switching valve. Plasma samples (10 μL) were injected directly onto a C18 SPE cartridge (MF Ph-1 C18, 10 mm × 4 mm, 5 μm) and the biological matrix was washed out for 2 min with the loading solvent (5 mM NaH(2)PO(4) buffer, pH 3.5) at a flow rate of 1 mL/min. By rotation of the switching valve, bavachinin was eluted from the SPE cartridge in the back-flush mode and transferred to the analytical column (Venusil MP C18, 4.6 mm × 150 mm, 5 μm) by the chromatographic mobile phase consisted of acetonitrile-5mM NaH(2)PO(4) buffer 65/35 (v/v, pH 3.5) at a flow rate of 1 mL/min. The complete cycle of the on-line SPE purification and chromatographic separation of the analyte was 13 min with UV detection performed at 236 nm. Calibration curve with good linearity (r=0.9997) was obtained in the range of 20-4000 ng/mL in mouse plasma. The intra-day and inter-day precisions (RSD) of bavachinin were in the range of 0.20-2.32% and the accuracies were between 98.47% and 102.95%. The lower limit of quantification (LLOQ) of the assay was 20 ng/mL. In conclusion, the established automated on-line SPE-HPLC-DAD method demonstrated good performance in terms of linearity, specificity, detection and quantification limits, precision and accuracy, and was successfully utilized to quantify bavachinin in mouse plasma to support the pharmacokinetic (PK) studies. The PK properties of bavachinin were characterized as rapid oral absorption, high clearance, and poor absolute bioavailability.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bavachinin, ≥95% (LC/MS-ELSD)
Prezzi e disponibilità al momento non sono disponibili