Merck
Pharmacological properties of lorglumide as a member of a new class of cholecystokinin antagonists.
Arzneimittel-Forschung (1987-11-01)
F Makovec, M Bani, R Cereda, R Chisté, M A Pacini, L Revel, L A Rovati, L C Rovati, I Setnikar
ABSTRACT
Derivatives of 5-(dipentylamino)-5-oxo-pentanoic acid are a new class of non-peptide cholecystokinin (CCK) antagonists. The most potent compound, D,L-4-(3,4-dichlorobenzoylamino)-5-(dipentylamino)-5-oxo-pen tanoic acid (lorglumide, CR 1409), has a great affinity for the pancreatic CCK receptors and is a competitive, specific and potent CCK antagonist on the smooth muscles of the gall bladder and ileum of the guinea pig and on the CCK-induced amylase secretion of isolated pancreatic acini. In vivo lorglumide antagonizes the contraction of the gall bladder of the guinea pig and of the dog provoked by i.v. CCK-8 or ceruletide (caerulein). It antagonizes the satiety effect of CCK-8 in the rat and is protective against ceruletide-, taurocholate- and diet-induced pancreatitis. Lorglumide is therefore a useful pharmacological tool to study the functions of CCK. For its pharmacological properties, its relatively low toxicity and because it is active also after oral administration, lorglumide is a candidate for diagnostic or therapeutic use in man when an involvement of CCK is suspected.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Lorglumide sodium salt, solid