Merck
Highly phosphomannosylated enzyme replacement therapy for GM2 gangliosidosis.
Annals of neurology (2011-04-27)
Daisuke Tsuji, Hiromi Akeboshi, Kazuhiko Matsuoka, Hiroko Yasuoka, Eri Miyasaki, Yoshiko Kasahara, Ikuo Kawashima, Yasunori Chiba, Yoshifumi Jigami, Takao Taki, Hitoshi Sakuraba, Kohji Itoh
ABSTRACT
Novel recombinant human lysosomal β-hexosaminidase A (HexA) was developed for enzyme replacement therapy (ERT) for Tay-Sachs and Sandhoff diseases, ie, autosomal recessive GM2 gangliosidoses, caused by HexA deficiency. A recombinant human HexA (Om4HexA) with a high mannose 6-phosphate (M6P)-type-N-glycan content, which was produced by a methylotrophic yeast strain, Ogataea minuta, overexpressing the OmMNN4 gene, was intracerebroventricularly (ICV) administered to Sandhoff disease model mice (Hexb⁻/⁻ mice) at different doses (0.5-2.5 mg/kg), and then the replacement and therapeutic effects were examined. The Om4HexA was widely distributed across the ependymal cell layer, dose-dependently restored the enzyme activity due to uptake via cell surface cation-independent M6P receptor (CI-M6PR) on neural cells, and reduced substrates, including GM2 ganglioside (GM2), asialo GM2 (GA2), and oligosaccharides with terminal N-acetylglucosamine residues (GlcNAc-oligosaccharides), accumulated in brain parenchyma. A significant inhibition of chemokine macrophage inflammatory protein-1 α (MIP-1α) induction was also revealed, especially in the hindbrain (< 63%). The decrease in central neural storage correlated with an improvement of motor dysfunction as well as prolongation of the lifespan. This lysosome-directed recombinant human enzyme drug derived from methylotrophic yeast has the high therapeutic potential to improve the motor dysfunction and quality of life of the lysosomal storage diseases (LSDs) patients with neurological manifestations. We emphasize the importance of neural cell surface M6P receptor as a delivery target of neural cell-directed enzyme replacement therapy (NCDERT) for neurodegenerative metabolic diseases.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Phosphomannose Isomerase from Escherichia coli, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥50 units/mg protein