Merck
The m6A demethylase FTO promotes keloid formation by up-regulating COL1A1.
Annals of translational medicine (2023-02-11)
Shuai Ren, Yingchang Ji, Mengmeng Wang, Maodong Ye, Lvdong Huang, Xiangna Cai
ABSTRACT
Keloid is a dermal fibrotic disease characterized by excessive proliferation of dermal fibroblasts and deposition of excessive collagen. N6-methyladenosine (m6A) plays a significant role in numerous physiological and pathological regulatory processes in the human body. Fat mass and obesity-associated protein (FTO) is one of the most essential m6A demethylases. However, whether FTO has a regulatory role in keloid development remains to be determined. In this study, we investigated the effects of the m6A demethylase FTO on keloid formation by performing hematoxylin and eosin (H&E) staining, m6A dot blotting, transwell migration experiment, and methylated RNA immunoprecipitation quantitative polymerase chain reaction (MeRIP-qPCR) tests, as well as real-time PCR (RT-PCR) and Western blot assays. The H&E staining indicated abnormal arrangement and proliferation of fibroblasts in the keloid tissue. The m6A dot blotting and qPCR revealed lower levels of m6A modification and increased expression of the m6A demethylases FTO in keloid tissue. Furthermore, overexpression of FTO promoted fibroblast migration as well as the expression of collagen type I alpha 1 chain (COL1A1) and α-smooth muscle actin (α-SMA). Mechanistic experiments demonstrated that FTO enhances keloid formation by modulating COL1A1 m6A modification and messenger RNA (mRNA) stability. In addition, this study also revealed the role of FTO in the therapeutic effect of glucocorticoids on keloids. Our study demonstrates that FTO upregulates COL1A1 expression via regulating COL1A1 m6A modification and maintaining mRNA stability, hence promoting keloid development and providing a potential new therapeutic target for the treatment of keloids.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-N6-Methyladenosine (m6A) antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM362, purified immunoglobulin