Merck
  • Down-regulation of Notch receptor signaling pathway induces caspase-dependent and caspase-independent apoptosis in lung squamous cell carcinoma cells.
Down-regulation of Notch receptor signaling pathway induces caspase-dependent and caspase-independent apoptosis in lung squamous cell carcinoma cells.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2012-05-16)
Huiqiu Cao, Yulin Hu, Peng Wang, Jianhua Zhou, Zhenghao Deng, Jifang Wen
ABSTRACT
Notch receptor signaling pathway (NRSP) is increasingly linked to carcinogenesis. Non-small cell lung cancer (NSCLC) appears to actively utilize this conserved developmental pathway. The aims of this study are to determine whether or not Notch 1-4 are overexpressed in NSCLC tissues compared with normal lung tissues and whether inhibiting NRSP could induce caspase-dependent or caspase-independent apoptosis. Immunohistochemistry was used to evaluate the expression of Notch 1-4 in 101 NSCLC tissue samples and 30 normal lung tissue samples. DAPT was used to repress NRSP in SK-MES-1 cells. Apoptosis was determined by Annexin V and PI staining. Cleaved poly ADP-ribose polymerase (PARP) was measured by Western blot; X-linked inhibitor of apoptosis protein (XIAP) and Survivin were assessed by qRT-PCR and Western blot; the release of second mitochondria-derived activator of caspase (Smac) from mitochondria to cytoplasm was evaluated by Western blot; the subcellular locations of endonuclease G (Endo G) and apoptosis inducing factor (AIF) were observed by Western blot and indirect immunofluorescence analysis. (Mech Dev, 98, 2000, 95) Notch 1-4 are up-regulated in NSCLC tissues and Notch 1, 2 are positively correlated with lymph node metastasis, (Proc Natl Acad Sci U S A, 106, 2009, 22293) DAPT treatment could inhibit NRSP and induce apoptosis, with a marked increase in cleaved PARP, decreases in XIAP and Survivin proteins and concomitant release of Smac, EndoG, and AIF from mitochondria, indicating that inhibiting NRSP by DAPT triggers caspase-dependent and caspase-independent apoptosis.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-DIABLO antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution