Merck
  • Simultaneous determination of pyridaben, dinotefuran, DN and UF in eggplant ecosystem under open-field conditions: Dissipation behaviour and residue distribution.
Simultaneous determination of pyridaben, dinotefuran, DN and UF in eggplant ecosystem under open-field conditions: Dissipation behaviour and residue distribution.
Chemosphere (2017-12-23)
Xiaoxin Chen, Xinze Liu, Bizhang Dong, Jiye Hu
ABSTRACT
A sensitive method for simultaneous determination of pyridaben, dinotefuran, DN and UF in eggplant ecosystem was established and validated through rapid resolution liquid chromatography triples quadrupole tandem mass spectrometry (RRLC-QqQ-MS/MS). Matrix-matched external calibrations were introduced to check matrix effects. Limits of quantification (LOQs) of pyridaben, dinotefuran, DN and UF in eggplant were 0.2, 0.2, 1.0 and 1.0 μg kg-1, and 0.2, 0.2, 5.0 and 1.0 μg kg-1 in soil, respectively. Limits of detection (LODs) of four pesticides were below 0.41 μg L-1. The mean recoveries (n = 5) of these insecticides varied from 79.4% to 103%, and the relative standard deviations (RSDs) ranged from 2.1% to 15.3% at three levels. This method was applied to Chinese open-field samples from two representative locations, which were previously treated with these insecticides at the doses of 210-315 g a.i. ha-1 twice or three times. The dissipations of pyridaben and dinotefuran in eggplant and soil followed first-order kinetics with the half-lives of 3.65-11.4 d. The residues of pyridaben and total dinotefuran (calculated as sum of dinotefuran parent, DN and UF) in eggplant were below 0.0311 mg kg-1 at the pre-harvest interval (PHI, 7 d). Presently, no maximum residue limit (MRL) of pyridaben and dinotefuran in eggplant was recommended by China, Codex Alimentarius Commission (CAC) or European Union (EU). This study was important for evaluation of environmental fate and food safety of pyridaben and dinotefuran in eggplant ecosystems, and facilitated China to establish maximum residue limits (MRLs) of pyridaben and dinotefuran in eggplant.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Supelco
Dinotefuran, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland