Merck
  • Contractile responses to endothelin-1 are regulated by PKC phosphorylation of cardiac myosin binding protein-C in rat ventricular myocytes.
Contractile responses to endothelin-1 are regulated by PKC phosphorylation of cardiac myosin binding protein-C in rat ventricular myocytes.
Journal of molecular and cellular cardiology (2018-02-23)
Ioannis Smyrnias, Normann Goodwin, Dagmar Wachten, Jonas Skogestad, Jan Magnus Aronsen, Emma L Robinson, Kateryna Demydenko, Anne Segonds-Pichon, David Oxley, Sakthivel Sadayappan, Karin Sipido, Martin D Bootman, H Llewelyn Roderick
ABSTRACT
The shortening of sarcomeres that co-ordinates the pump function of the heart is stimulated by electrically-mediated increases in [Ca2+]. This process of excitation-contraction coupling (ECC) is subject to modulation by neurohormonal mediators that tune the output of the heart to meet the needs of the organism. Endothelin-1 (ET-1) is a potent modulator of cardiac function with effects on contraction amplitude, chronotropy and automaticity. The actions of ET-1 are evident during normal adaptive physiological responses and increased under pathophysiological conditions, such as following myocardial infarction and during heart failure, where ET-1 levels are elevated. In myocytes, ET-1 acts through ETA- or ETB-G protein-coupled receptors (GPCRs). Although well studied in atrial myocytes, the influence and mechanisms of action of ET-1 upon ECC in ventricular myocytes are not fully resolved. We show in rat ventricular myocytes that ET-1 elicits a biphasic effect on fractional shortening (initial transient negative and sustained positive inotropy) and increases the peak amplitude of systolic Ca2+ transients in adult rat ventricular myocytes. The negative inotropic phase was ETB receptor-dependent, whereas the positive inotropic response and increase in peak amplitude of systolic Ca2+ transients required ETA receptor engagement. Both effects of ET-1 required phospholipase C (PLC)-activity, although distinct signalling pathways downstream of PLC elicited the effects of each ET receptor. The negative inotropic response involved inositol 1,4,5-trisphosphate (InsP3) signalling and protein kinase C epsilon (PKCε). The positive inotropic action and the enhancement in Ca2+ transient amplitude induced by ET-1 were independent of InsP3 signalling, but suppressed by PKCε. Serine 302 in cardiac myosin binding protein-C was identified as a PKCε substrate that when phosphorylated contributed to the suppression of contraction and Ca2+ transients by PKCε following ET-1 stimulation. Thus, our data provide a new role and mechanism of action for InsP3 and PKCε in mediating the negative inotropic response and in restraining the positive inotropy and enhancement in Ca2+ transients following ET-1 stimulation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-α-Actinin (Sarcomeric) antibody, Mouse monoclonal, clone EA-53, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-PKCepsilon Antibody (Ser729), from rabbit, purified by affinity chromatography