Merck
  • Interferon Regulatory Factor 4 Regulates the Development of Polymorphonuclear Myeloid-Derived Suppressor Cells Through the Transcription of c-Myc in Cancer.
Interferon Regulatory Factor 4 Regulates the Development of Polymorphonuclear Myeloid-Derived Suppressor Cells Through the Transcription of c-Myc in Cancer.
Frontiers in immunology (2021-03-13)
Quan Yang, Hongyan Xie, Xing Li, Yuanfa Feng, Shihao Xie, Jiale Qu, Anqi Xie, Yiqiang Zhu, Lu Zhou, Jinxue Yang, Xiaohao Hu, Haixia Wei, Huaina Qiu, Wenjuan Qin, Jun Huang
ABSTRACT
The accumulation of myeloid-derived suppressor cells (MDSCs) is one of the major obstacles to achieve an appropriate anti-tumor immune response and successful tumor immunotherapy. MDSCs in tumor-bearing hosts are primarily polymorphonuclear (PMN-MDSCs). However, the mechanisms regulating the development of MDSCs remain poorly understood. In this report, we showed that interferon regulatory factor 4 (IRF4) plays a key role in the development of PMN-MDSCs, but not monocytic MDSCs. IRF4 deficiency caused a significant elevation of PMN-MDSCs and enhanced the suppressive activity of PMN-MDSCs, increasing tumor growth and metastasis in mice. Mechanistic studies showed that c-Myc was up-regulated by the IRF4 protein. Over-expression of c-Myc almost abrogated the effects of IRF4 deletion on PMN-MDSCs development. Importantly, the IRF4 expression level was negatively correlated with the PMN-MDSCs frequency and tumor development but positively correlated with c-Myc expression in clinical cancer patients. In summary, this study demonstrated that IRF4 represents a novel regulator of PMN-MDSCs development in cancer, which may have predictive value for tumor progression.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
6-Carboxyfluorescein diacetate, ≥95% (HPLC)
Sigma-Aldrich
10074-G5, ≥98% (HPLC)