Merck
  • Toward establishing structure-activity relationships for oxygenated coumarins as differentiation inducers of promonocytic leukemic cells.
Toward establishing structure-activity relationships for oxygenated coumarins as differentiation inducers of promonocytic leukemic cells.
Bioorganic & medicinal chemistry (2009-09-01)
María E Riveiro, Dominick Maes, Ramiro Vázquez, Monica Vermeulen, Sven Mangelinckx, Jan Jacobs, Silvia Debenedetti, Carina Shayo, Norbert De Kimpe, Carlos Davio
ABSTRACT
The presumption that some coumarins might be lead compounds in the search for new differentiation agents against leukemia is based on the fact that natural coumarins, 5-(3-methyl-2-butenyloxy)-6,7-methylenedioxycoumarin (C-2) and 5-methoxy-6,7-methylenedioxycoumarin (C-1) inhibit proliferation and induce differentiation in U-937 cells [Riveiro, M. E.; Shayo, C.; Monczor, F.; Fernandez, N.; Baldi, A.; De Kimpe, N.; Rossi, J.; Debenedetti, S.; Davio, C. Cancer Lett.2004, 210, 179-188]. These promising findings prompted us to investigate the anti-leukemia activity of a broader range of related polyoxygenated coumarins. Twenty related natural or synthetically prepared coumarins, including a range of 5-substituted ayapin derivatives which have become easy accessible via newly developed synthesis methods, were evaluated, where treatments with 5-(2,3-dihydroxy-3-methylbutoxy)-6,7-methylenedioxycoumarin (D-3) and 5-(2-hydroxy-3-methoxy-3-methylbutoxy)-6,7-methylenedioxycoumarin (D-2) were able to inhibit the cell growth and induce the differentiation of U-937 cells after 48 h treatment. These results provide insight into the correlation between some structural properties of polyoxygenated coumarins and their in vitro leukemic differentiation activity.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Coumarin, ≥99% (HPLC)