Merck
  • Effect of granulocyte-macrophage colony-stimulating factor inducer on left ventricular remodeling after acute myocardial infarction.
Effect of granulocyte-macrophage colony-stimulating factor inducer on left ventricular remodeling after acute myocardial infarction.
Journal of the American College of Cardiology (2004-10-07)
Yuichiro Maekawa, Toshihisa Anzai, Tsutomu Yoshikawa, Yasuo Sugano, Keitaro Mahara, Takashi Kohno, Toshiyuki Takahashi, Satoshi Ogawa
ABSTRACT
We sought to determine the influence of granulocyte-macrophage colony-stimulating factor (GM-CSF) induction on post-myocardial infarction (MI) remodeling, especially in relation to the inflammatory response and myocardial fibrosis. Granulocyte-macrophage colony-stimulating factor modifies wound healing by promoting monocytopoiesis and infiltration of monocytes and macrophages into injured tissue; however, the effect of GM-CSF induction on the infarct healing process and myocardial fibrosis is unclear. A model of MI was produced in Wistar rats by ligation of the left coronary artery. The MI animals were randomized to receive GM-CSF inducer (romurtide 200 microg/kg/day for 7 consecutive days) (MI/Ro) or saline (MI/C). Echocardiographic and hemodynamic studies on day 14 revealed increased left ventricular (LV) end-diastolic dimension, decreased fractional shortening, elevated LV end-diastolic pressure, and decreased LV maximum rate of isovolumic pressure development in MI/Ro compared with MI/C. Immunoblotting showed that expression of transforming growth factor (TGF)-beta1 in the infarcted site on day 3 after MI was decreased in MI/Ro compared with MI/C. In the infarcted site, TGF-beta1, collagen type I and type III messenger ribonucleic acid (mRNA) expression on day 3, and collagen content on day 7 were reduced in MI/Ro compared with MI/C, in association with marked infarct expansion. In MI/Ro, monocyte chemoattractant protein-1 mRNA level and the degree of infiltration of monocyte-derived macrophages (ED-1-positive)were greater in the infarcted site on day 7 than those in MI/C. The GM-CSF induction by romurtide facilitated infarct expansion in association with the promotion of monocyte recruitment and inappropriate collagen synthesis in the infarcted region during the early phase of MI.