Merck
TNF-α in T lymphocytes attenuates renal injury and fibrosis during nephrotoxic nephritis.
American journal of physiology. Renal physiology (2019-11-19)
Yi Wen, Nathan P Rudemiller, Jiandong Zhang, Taylor Robinette, Xiaohan Lu, Jiafa Ren, Jamie R Privratsky, Sergei A Nedospasov, Steven D Crowley
ABSTRACT
Nephrotoxic serum nephritis (NTN) models immune-mediated human glomerulonephritis and culminates in kidney inflammation and fibrosis, a process regulated by T lymphocytes. TNF-α is a key proinflammatory cytokine that contributes to diverse forms of renal injury. Therefore, we posited that TNF-α from T lymphocytes may contribute to NTN pathogenesis. Here, mice with T cell-specific deletion of TNF-α (TNF TKO) and wild-type (WT) control mice were subjected to the NTN model. At 14 days after NTN, kidney injury and fibrosis were increased in kidneys from TNF TKO mice compared with WT mice. PD1+CD4+ T cell numbers and mRNA levels of IL-17A were elevated in NTN kidneys of TNF TKO mice, suggesting that augmented local T helper 17 lymphocyte responses in the TNF TKO kidney may exaggerate renal injury and fibrosis. In turn, we found increased accumulation of neutrophils in TNF TKO kidneys during NTN. We conclude that TNF-α production in T lymphocytes mitigates NTN-induced kidney injury and fibrosis by inhibiting renal T helper 17 lymphocyte responses and infiltration of neutrophils.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-Sheep IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution