Merck
Fatty acids derived from royal jelly are modulators of estrogen receptor functions.
PloS one (2011-01-05)
Paraskevi Moutsatsou, Zoi Papoutsi, Eva Kassi, Nina Heldring, Chunyan Zhao, Anna Tsiapara, Eleni Melliou, George P Chrousos, Ioanna Chinou, Andrey Karshikoff, Lennart Nilsson, Karin Dahlman-Wright
ABSTRACT
Royal jelly (RJ) excreted by honeybees and used as a nutritional and medicinal agent has estrogen-like effects, yet the compounds mediating these effects remain unidentified. The possible effects of three RJ fatty acids (FAs) (10-hydroxy-2-decenoic-10H2DA, 3,10-dihydroxydecanoic-3,10DDA, sebacic acid-SA) on estrogen signaling was investigated in various cellular systems. In MCF-7 cells, FAs, in absence of estradiol (E(2)), modulated the estrogen receptor (ER) recruitment to the pS2 promoter and pS2 mRNA levels via only ERβ but not ERα, while in presence of E(2) FAs modulated both ERβ and ERα. Moreover, in presence of FAs, the E(2)-induced recruitment of the EAB1 co-activator peptide to ERα is masked and the E(2)-induced estrogen response element (ERE)-mediated transactivation is inhibited. In HeLa cells, in absence of E(2), FAs inhibited the ERE-mediated transactivation by ERβ but not ERα, while in presence of E(2), FAs inhibited ERE-activity by both ERβ and ERα. Molecular modeling revealed favorable binding of FAs to ERα at the co-activator-binding site, while binding assays showed that FAs did not bind to the ligand-binding pocket of ERα or ERβ. In KS483 osteoblasts, FAs, like E(2), induced mineralization via an ER-dependent way. Our data propose a possible molecular mechanism for the estrogenic activities of RJ's components which, although structurally entirely different from E(2), mediate estrogen signaling, at least in part, by modulating the recruitment of ERα, ERβ and co-activators to target genes.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Reagente per trasfezione GeneJuice®, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.