Merck
  • Synthesis, spectral analysis and biological evaluation of sulfamoyl and 1,3,4-oxadiazole derivatives of 3-pipecoline.
Synthesis, spectral analysis and biological evaluation of sulfamoyl and 1,3,4-oxadiazole derivatives of 3-pipecoline.
Pakistan journal of pharmaceutical sciences (2019-07-07)
Azizur Rehman, Saira Jabeen Aslam, Muhammad Athar Abbasi, Sabahat Zahra Siddiqui, Shahid Rasool, Syed Adnan Ali Shah
ABSTRACT
Heterocyclic chemistry is an important field of organic chemistry due to therapeutic potential. The minor modification in the structure of poly-functional compounds has great effect on therapeutic ability. In the presented research work, substituted 1,3,4-oxadiazole derivatives, 8a-p, have been synthesized by the reaction of 1-(4-bromomethylbenzenesulfonyl)-3-methylpiperidine (7) and 5-substituted-1,3,4-oxadiazole-2-thiol (4a-p). The 5-substituted-1,3,4-oxadiazole-2-thiol were synthesized by converting carboxylic acids correspondingly into esters, hydrazides and oxadiazoles. Secondly the electrophile, 1-(4-Bromomethylbenzenesulfonyl)-3-methylpiperidine (7), was prepared by the reaction of 3-methylpiperidine with 4-bromomethylbenzenesulfonyl chloride in the presence of water and Na2CO3 under pH of 9-10. The compounds were structurally corroborated through spectroscopic data analysis of IR, EI-MS and 1H-NMR. The screening for antibacterial activity revealed the compounds to be moderate to excellent inhibitors against bacteria under study. Anti-enzymatic activity was assessed against urease enzyme and 1-{[4-({[5-(3-nitrophenyl)-1,3,4-oxadiazol-2-yl]sulfanyl}methyl)phenyl]sulfonyl}-3-methylpiperidine (8d) was the most active one.