Merck
  • Metabolomic association between venous thromboembolism in critically ill trauma patients and kynurenine pathway of tryptophan metabolism.
Metabolomic association between venous thromboembolism in critically ill trauma patients and kynurenine pathway of tryptophan metabolism.
Thrombosis research (2018-03-16)
Stacy A Voils, Mohamed H Shahin, Timothy J Garrett, Reginald F Frye
ABSTRACT
Incidence of venous thromboembolism (VTE) in critically ill patients remains unacceptably high despite widespread use of thromboprophylaxis. A systems biology approach may be useful in understanding disease pathology and predicting response to treatment. Metabolite profile under specific environmental conditions provides the closest link to phenotype, but the relationship between metabolomics and risk of VTE in critically ill patients is unknown. In this study, metabolomics signatures are compared in patients with and without VTE. Multicenter case-control study using prospectively collected data from the Inflammation and Host Response to Injury program, with pathway and in silico gene expression analyses. Eight level 1 US trauma centers. Critically ill adults with blunt trauma who developed VTE within the first 28 days of hospitalization compared to patients without VTE (N-VTE). None. Patients included in the study (n = 20 VTE, n = 20 N-VTE) were mean age of 34 years, injury severity score of 35, and VTE diagnosed a median of 10.5 days after admission. Global metabolomics revealed two kynurenine metabolites, N-formylkynurenine (AUC = 0.77; 95% CI: 0.59-0.89) and 5-hydroxy-N-formylkynurenine (AUC = 0.80; 95% CI:0.63-0.90) significantly discriminated VTE and N-VTE; ratio between N-formylkynurenine/5-hydroxy-N-formylkynurenine improved predictive power (AUC = 0.87; 95% CI: 0.74-0.95). In the pathway analysis, tryptophan was the only significant metabolic pathway including N-formylkynurenine and 5-hydroxy-N-formylkynurenine (p < 0.001), and 8 proteins directly or indirectly interacted with these metabolites in the interaction network analysis. Of the 8 genes tested in the in silico gene expression analyses, KYNU (p < 0.001), CCBL1 (p < 0.001), and CCBL2 (p = 0.001) were significantly different between VTE and N-VTE, controlling for age and sex. Two novel kynurenine metabolites in the tryptophan pathway associated with hospital-acquired VTE, and 3 candidate genes were identified via pathway and interaction network analyses. Future studies are warranted to validate these findings in diverse populations using a multi-omics approach.