Merck
  • Reversible Addition?Fragmentation Chain-Transfer Polymerization of Amphiphilic Polycarboxybetaines and Their Molecular Interactions.
Reversible Addition?Fragmentation Chain-Transfer Polymerization of Amphiphilic Polycarboxybetaines and Their Molecular Interactions.
Langmuir : the ACS journal of surfaces and colloids (2019-06-15)
Munziya Abutalip, Anam Mahmood, Raikhan Rakhmetullayeva, Alexey Shakhvorostov, Yerbol Dauletov, Sarkyt Kudaibergenov, Nurxat Nuraje
ABSTRACT
In this work, we report the first molecular weight-controlled amphiphilic polybetaine synthesis using various hydrocarbons via reversible addition?fragmentation chain-transfer (RAFT) polymerization. The experimental separation of the alkyl aminocrotonate tautomers, which has been the subject of debate, was completed for the first time. The enamine form of these tautomers was further used as a monomer for the RAFT polymerization of amphiphilic polycarboxybetaines. Self-assembly of the amphiphilic polycarboxybetaines showed micelle structures from spherical, rod-like to fractal in the aqueous media due to the competition between both electrostatic and hydrophobic forces. Hydrophobically dominant interactions among amphiphilic polycarboxybetaines and long-chain hydrocarbon alkane molecules were investigated to understand long-chain hydrocarbon alkane crystallization using alkane crystal deposition and viscosity experiments. Strong hydrophobic forces between poly(hexadecyl-grafted aminocrotonate?methacrylic acid) and long-chain hydrocarbon alkane molecules changed the surface properties of the long-chain hydrocarbon alkane nucleus and inhibited the growth of paraffin crystals.