Merck
The pharmacologic characterization of allosteric molecules: Gq protein activation.
Journal of receptor and signal transduction research (2019-07-20)
Sara Bdioui, Julien Verdi, Nicolas Pierre, Eric Trinquet, Thomas Roux, Terry Kenakin
ABSTRACT
Context: Drugs such as positive allosteric modulators (PAMs) produce complex behaviors when acting on tissues in different physiological contexts in vivo. Objective: This study describes the use of functional assays of varying receptor sensitivity to unveil the various behaviors of PAMs and thus quantify allosteric effect through system independent scales. Materials and methods: Muscarinic receptor activation with acetylcholine (ACh) was used to the demonstrate activity of the PAM agonist 1-(4-methoxybenzyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, Benzyl quinolone carboxylic acid (BQCA) in terms of direct agonism, potentiation of ACh affinity, and ACh efficacy. Concentration-response curves were fit to the functional allosteric model to yield indices of agonism (τB), effects on affinity (α cooperativity), and efficacy (β cooperativity). Results: It is shown that a highly sensitive functional assay revealed the direct efficacy of BQCA as an agonist and relatively insensitive cells (produced by chemical alkylation of muscarinic receptor with phenoxybenzamine) revealed a positive allosteric effect of BQCA on ACh efficacy. A wide range of functional assay sensitivities produced a complex pattern of behavior for BQCA all of which was accurately quantified through the system-independent parameters of the functional allosteric model. Conclusions: The study of complex allosteric molecules in a range of functional assays of varying sensitivity allows the measurement of the complete array of activities of these molecules on receptors and also better predicts which will be seen with these in vivo where a range of tissue sensitivities is encountered.