Merck
Identification of non-volatile migrants from baby bottles by UPLC-Q-TOF-MS.
Food research international (Ottawa, Ont.) (2019-07-10)
Wellington da Silva Oliveira, Sara Ubeda, Cristina Nerín, Marisa Padula, Helena Teixeira Godoy
ABSTRACT
Baby bottles made of polypropylene, Tritan® and silicone were evaluated regarding the migration of non-volatile compounds using UPLC-QTOF-MS. Twenty-seven compounds were identified. In all polypropylene samples the migration of 2.2'-(tridecylimino)bis-ethanol and derivatives thereof were detected in concentrations below the specific migration limit (1.2 mg.kg-1). Furthermore, clarifying agents and glycerol derivatives were detected. Tritan baby bottle showed the migration of one slip additive. On the other hand, twenty compounds were detected in silicone baby bottles. Most of them were unknown compounds derived from acrylates. Once the migrants were identified, the risk assessment was carried out using the Threshold of Toxicological Concern (TTC) approach. The risk assessment of migrants coming from silicone samples showed levels above the threshold recommended as safe for babies.