Merck
  • Hu Antigen R Regulates Antiviral Innate Immune Responses through the Stabilization of mRNA for Polo-like Kinase 2.
Hu Antigen R Regulates Antiviral Innate Immune Responses through the Stabilization of mRNA for Polo-like Kinase 2.
Journal of immunology (Baltimore, Md. : 1950) (2018-04-22)
Takuya Sueyoshi, Takumi Kawasaki, Yuichi Kitai, Daisuke Ori, Shizuo Akira, Taro Kawai
ABSTRACT
Retinoic acid-inducible gene I (RIG-I)-like receptors (RLRs), RIG-I, and melanoma differentiation-associated gene 5 (MDA5) play a critical role in inducing antiviral innate immune responses by activating IFN regulatory factor 3 (IRF3) and NF-κB, which regulates the transcription of type I IFN and inflammatory cytokines. Antiviral innate immune responses are also regulated by posttranscriptional and translational mechanisms. In this study, we identified an RNA-binding protein HuR as a regulator for RLR signaling. Overexpression of HuR, but not of other Hu members, increased IFN-β promoter activity. HuR-deficient macrophage cells exhibited decreased Ifnb1 expression after RLR stimulation, whereas they showed normal induction after stimulation with bacterial LPS or immunostimulatory DNA. Moreover, HuR-deficient cells displayed impaired nuclear translocation of IRF3 after RLR stimulation. In HuR-deficient cells, the mRNA expression of Polo-like kinase (PLK) 2 was markedly reduced. We found that HuR bound to the 3' untranslated region of Plk2 mRNA and increased its stabilization. PLK2-deficient cells also showed reduced IRF3 nuclear translocation and Ifnb mRNA expression during RLR signaling. Together, these findings suggest that HuR bolsters RLR-mediated IRF3 nuclear translocation by controlling the stability of Plk2 mRNA.