Merck
  • Inhibition of VEGF (Vascular Endothelial Growth Factor)-A or its Receptor Activity Suppresses Experimental Aneurysm Progression in the Aortic Elastase Infusion Model.
Inhibition of VEGF (Vascular Endothelial Growth Factor)-A or its Receptor Activity Suppresses Experimental Aneurysm Progression in the Aortic Elastase Infusion Model.
Arteriosclerosis, thrombosis, and vascular biology (2019-07-12)
Baohui Xu, Yasunori Iida, Keith J Glover, Yingbin Ge, Yan Wang, Haojun Xuan, Xiaolei Hu, Hiroki Tanaka, Wei Wang, Naoki Fujimura, Masaaki Miyata, Takahiro Shoji, Jia Guo, Xiaoya Zheng, Mary Gerritsen, Calvin Kuo, Sara A Michie, Ronald L Dalman
ABSTRACT
We examined the pathogenic significance of VEGF (vascular endothelial growth factor)-A in experimental abdominal aortic aneurysms (AAAs) and the translational value of pharmacological VEGF-A or its receptor inhibition in aneurysm suppression. Approaches and Results: AAAs were created in male C57BL/6J mice via intra-aortic elastase infusion. Soluble VEGFR (VEGF receptor)-2 extracellular ligand-binding domain (delivered in Ad [adenovirus]-VEGFR-2), anti-VEGF-A mAb (monoclonal antibody), and sunitinib were used to sequester VEGF-A, neutralize VEGF-A, and inhibit receptor tyrosine kinase activity, respectively. Influences on AAAs were assessed using ultrasonography and histopathology. In vitro transwell migration and quantitative reverse transcription polymerase chain reaction assays were used to assess myeloid cell chemotaxis and mRNA expression, respectively. Abundant VEGF-A mRNA and VEGF-A-positive cells were present in aneurysmal aortae. Sequestration of VEGF-A by Ad-VEGFR-2 prevented AAA formation, with attenuation of medial elastolysis and smooth muscle depletion, mural angiogenesis and monocyte/macrophage infiltration. Treatment with anti-VEGF-A mAb prevented AAA formation without affecting further progression of established AAAs. Sunitinib therapy substantially mitigated both AAA formation and further progression of established AAAs, attenuated aneurysmal aortic MMP2 (matrix metalloproteinase) and MMP9 protein expression, inhibited inflammatory monocyte and neutrophil chemotaxis to VEGF-A, and reduced MMP2, MMP9, and VEGF-A mRNA expression in macrophages and smooth muscle cells in vitro. Additionally, sunitinib treatment reduced circulating monocytes in aneurysmal mice. VEGF-A and its receptors contribute to experimental AAA formation by suppressing mural angiogenesis, MMP and VEGF-A production, myeloid cell chemotaxis, and circulating monocytes. Pharmacological inhibition of receptor tyrosine kinases by sunitinib or related compounds may provide novel opportunities for clinical aneurysm suppression.