Merck
  • Chondroitin sulfate-D promotes neurite outgrowth by acting as an extracellular ligand for neuronal integrin αVβ3.
Chondroitin sulfate-D promotes neurite outgrowth by acting as an extracellular ligand for neuronal integrin αVβ3.
Biochimica et biophysica acta. General subjects (2019-06-11)
Miharu Shida, Tadahisa Mikami, Jun-Ichi Tamura, Hiroshi Kitagawa
ABSTRACT
Chondroitin sulfate (CS) chains are prominent extra/pericellular matrix components in the central nervous system (CNS) and can exert positive or negative regulatory effects on neurite outgrowth, depending on the CS structure and the amount. Despite the remarkable abilities of highly sulfated forms of CS chains to enhance neurite outgrowth, the neuronal recognition systems for such promotional CS chains, including CS-D polysaccharide, remain to be fully elucidated. We explored the molecular basis of the CS-D-mediated neurite extension using primary hippocampal neurons cultured on substrate precoated with CS-D polysaccharides, and evaluated functional involvement of a distinct integrin heterodimer as a novel neuronal CS receptor for CS-D. We identified an extracellular matrix receptor, integrin αVβ3, as a functional receptor for CS-D. CS-D, but not CS-C (a precursor form of CS-D) showed significant binding affinity toward recombinant integrin αVβ3 heterodimer and activated intracellular signaling(s) involving focal adhesion kinase (FAK) and Src/Fyn kinase. Functional blockade of the respective players for integrin signaling abrogated the promotional effects of CS-D. We also found the existence of CS-D-induced integrin activation system in neuronal stem/progenitor cell population. The neuronal cell surface integrin αVβ3 can function as a CS receptor for a highly sulfated CS subtype, CS-D. Our findings are the first to demonstrate that CS-dependent neurite outgrowth promotion is exerted via direct activation of specific integrin heterodimers on neuronal cell surfaces, providing new insights into understanding the CS-sensing machineries that regulate CNS development and regeneration.