Merck
A novel form of JARID2 is required for differentiation in lineage-committed cells.
The EMBO journal (2018-12-24)
Diaa Al-Raawi, Rhian Jones, Susanne Wijesinghe, John Halsall, Marija Petric, Sally Roberts, Neil A Hotchin, Aditi Kanhere
ABSTRACT
Polycomb repressive complex-2 (PRC2) is a group of proteins that play an important role during development and in cell differentiation. PRC2 is a histone-modifying complex that catalyses methylation of lysine 27 of histone H3 (H3K27me3) at differentiation genes leading to their transcriptional repression. JARID2 is a co-factor of PRC2 and is important for targeting PRC2 to chromatin. Here, we show that, unlike in embryonic stem cells, in lineage-committed human cells, including human epidermal keratinocytes, JARID2 predominantly exists as a novel low molecular weight form, which lacks the N-terminal PRC2-interacting domain (ΔN-JARID2). We show that ΔN-JARID2 is a cleaved product of full-length JARID2 spanning the C-terminal conserved jumonji domains. JARID2 knockout in keratinocytes results in up-regulation of cell cycle genes and repression of many epidermal differentiation genes. Surprisingly, repression of epidermal differentiation genes in JARID2-null keratinocytes can be rescued by expression of ΔN-JARID2 suggesting that, in contrast to PRC2, ΔN-JARID2 promotes activation of differentiation genes. We propose that a switch from expression of full-length JARID2 to ΔN-JARID2 is important for the up-regulation differentiation genes.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-trimetil-istone H3 (Lys27), Upstate®, from rabbit