Merck
  • Alginate lyase immobilized chitosan nanoparticles of ciprofloxacin for the improved antimicrobial activity against the biofilm associated mucoid P. aeruginosa infection in cystic fibrosis.
Alginate lyase immobilized chitosan nanoparticles of ciprofloxacin for the improved antimicrobial activity against the biofilm associated mucoid P. aeruginosa infection in cystic fibrosis.
International journal of pharmaceutics (2019-03-31)
Krishna Kumar Patel, Muktanand Tripathi, Nidhi Pandey, Ashish Kumar Agrawal, Shilpkala Gade, Md Meraj Anjum, Ragini Tilak, Sanjay Singh
ABSTRACT
Dense colonization of mucoid Pseudomonas aeruginosa within the self-secreted extracellular matrix (mainly alginate), called biofilm, is a principal reason for the failure of antimicrobial therapy in cystic fibrotic patients. Alginate is a key component in the biofilm of mucoid P. aeruginosa and responsible for surface adhesion and stabilization of biofilm. To overcome this problem, alginate lyase functionalized chitosan nanoparticles of ciprofloxacin were developed for the effective treatment of P. aeruginosa infection in cystic fibrosis patients. The developed nanoparticles were found to have desired quality attributes and demonstrated sustained release following the Higuchi release kinetics. Drug compatibility with the chitosan was confirmed by FTIR while powder X-ray diffraction analysis confirmed the entrapment of drug within the nanoparticle matrix. Lactose adsorbed NPs showed promising aerodynamic property. Nanoparticles showed prolonged MIC and significant reduction in biofilm aggregation and formation in planktonic bacterial suspension. Nanoparticles exhibited significantly higher inhibitory effect against biofilm of P. aeruginosa and reduced the biomass, thickness and density confirmed by confocal microscopy. Furthermore, developed nanoparticles were haemocompatible and did not exhibit any toxicity in vitro MTT assay and in vivo on lungs male Wistar rats. The data in hand collectively suggest the proposed strategy a better alternative for the effective treatment of cystic fibrosis infections.