Merck
  • Evaluation of the Effects of a Monthly Buprenorphine Depot Subcutaneous Injection on QT Interval During Treatment for Opioid Use Disorder.
Evaluation of the Effects of a Monthly Buprenorphine Depot Subcutaneous Injection on QT Interval During Treatment for Opioid Use Disorder.
Clinical pharmacology and therapeutics (2019-02-26)
Virginia D Schmith, Laura Curd, Lauren R L Lohmer, Celine M Laffont, Anne Andorn, Malcolm A Young
ABSTRACT
Extensive 12-lead electrocardiogram monitoring and drug concentrations were obtained during development of BUP-XR, a monthly subcutaneous injection for the treatment of opioid use disorder (OUD). Matched QT and plasma drug concentrations (11,925) from 1,114 subjects were pooled from 5 studies in OUD. A concentration-QT model was developed, which accounted for confounding factors (e.g., comedications) affecting heart rate and heart rate-corrected QT interval (QTc). Bias-corrected nonparametric two-sided 90% confidence intervals (CIs) were derived for the mean predicted effect of BUP-XR on QTc (ΔQTc) at therapeutic and supratherapeutic doses. Changes in QTc were associated with age, central vs. noncentral reading, sex, methadone, and barbiturates. The upper 90% CI of ΔQTc was 0.29, 0.67, and 1.34 ms at the steady-state peak concentration (Cmax ) for 100, 300, and 2 × 300 mg doses, respectively. An effect of BUP-XR on QT can be ruled out at therapeutic and supratherapeutic doses of BUP-XR, after accounting for covariates that may influence heart rate and QT interval in OUD.