Merck
  • Ras/Raf1-dependent signal in sphingosine-1-phosphate-induced tube formation in human coronary artery endothelial cells.
Ras/Raf1-dependent signal in sphingosine-1-phosphate-induced tube formation in human coronary artery endothelial cells.
Biochemical and biophysical research communications (2003-06-25)
Shin-ichiro Miura, Hiroyuki Tanigawa, Yoshino Matsuo, Masahiro Fujino, Akira Kawamura, Keijiro Saku
ABSTRACT
Since we recently reported that high density lipoprotein, which contains the bioactive lipid sphingosine-1-phosphate (S1P) [Arterioscler. Thromb. Vasc. Biol. 23 (2003) 802], induced human coronary artery endothelial cell (HCEC) tube formation mediated by a Ras/Raf/ERK (extracellular signal-activated kinase) pathway, we thought that it would be very important to evaluate whether the signal in S1P-induced tube formation is Ras-dependent or -independent. In an in vitro model of HCEC tube formation on a matrix gel, S1P-induced tube formation. ERK1/2 inhibitor (PD98059) and pertussis toxin (PTX) suppressed S1P-induced tube formation. S1P activated phospho(p)-ERK1/2, while dominant-negative RasN17 blocked S1P-induced p-ERK1/2. Moreover, RasN17 inhibited S1P-induced tube formation. S1P activated Ras/Raf1 by Ras pull-down assay and this effect was inhibited by PTX. These results demonstrate that Ras/Raf1-dependent ERK activation mediated by PTX-sensitive G protein-coupled receptors may be a potent signal in S1P-induced HCEC tube formation.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Kit per il dosaggio dell'attivazione di RAS