Merck
BACE1 inhibitory effects of lavandulyl flavanones from Sophora flavescens.
Bioorganic & medicinal chemistry (2008-06-21)
Eun Mi Hwang, Young Bae Ryu, Hoi Young Kim, Dong-Gyu Kim, Seong-Geun Hong, Jin Hwan Lee, Marcus J Curtis-Long, Seong Hun Jeong, Jae-Yong Park, Ki Hun Park
ABSTRACT
In order to access beta-secretase (BACE1), and enzyme strongly implicated in the cause of Alzheimer's disease, inhibitors must possess sufficient lipophilicity to traverse two lipid bilayers. Current drug candidates, which are almost totally peptide-derived, are thus inefficient because cell permeability presents a serious limiting factor. In this study, lipophilic alkylated (C(10)-C(5)) flavanones from Sophora flavescens were examined for their inhibitory effects against beta-secretase. Lavandulyl flavanones (1, 2, 5, 6, and 8) showed potent beta-secretase inhibitory activities with IC(50)s of 5.2, 3.3, 8.4, 2.6, and 6.7microM, respectively, while no significant activity was observed in the corresponding hydrated lavandulyl flavanones (4 and 7) and prenylated flavanone (3). As we expected, lavandulyl flavanones reduced Abeta secretion dose-dependently in transfected human embryonic kidney (HEK-293) cells. In kinetic studies, all compounds screened were shown to be noncompetitive inhibitor.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-APP Antibody, APP 643-695 CT fragment, clone 2.F2.19B4, ascites fluid, clone 2.F2.19B4, Chemicon®