Merck
  • Low Temperature Synthesis of Superparamagnetic Iron Oxide (Fe3O4) Nanoparticles and Their ROS Mediated Inhibition of Biofilm Formed by Food-Associated Bacteria.
Low Temperature Synthesis of Superparamagnetic Iron Oxide (Fe3O4) Nanoparticles and Their ROS Mediated Inhibition of Biofilm Formed by Food-Associated Bacteria.
Frontiers in microbiology (2018-11-23)
Nasser A Al-Shabib, Fohad Mabood Husain, Faheem Ahmed, Rais Ahmad Khan, Mohammad Shavez Khan, Firoz Ahmad Ansari, Mohammad Zubair Alam, Mohammed Asif Ahmed, Mohd Shahnawaz Khan, Mohammad Hassan Baig, Javed Masood Khan, Syed Ali Shahzad, Mohammed Arshad, Abdullah Alyousef, Iqbal Ahmad
ABSTRACT
In the present study, a facile environmentally friendly approach was described to prepare monodisperse iron oxide (Fe3O4) nanoparticles (IONPs) by low temperature solution route. The synthesized nanoparticles were characterized using x-ray diffraction spectroscopy (XRD), Raman spectroscopy, field emission scanning electron microscopy (FESEM) measurements, Fourier-Transform Infrared Spectroscopy (FTIR), and Thermogravimetric analysis (TGA) analyses. XRD patterns revealed high crystalline quality of the nanoparticles. SEM micrographs showed the monodispersed IONPs with size ranging from 6 to 9 nm. Synthesized nanoparticles demonstrated MICs of 32, 64, and 128 μg/ml against Gram negative bacteria i.e., Serratia marcescens, Escherichia coli, and Pseudomonas aeruginosa, respectively, and 32 μg/ml against Gram positive bacteria Listeria monocytogenes. IOPNs at its respective sub-MICs demonstrated significant reduction of alginate and exopolysaccharide production and subsequently demonstrated broad-spectrum inhibition of biofilm ranging from 16 to 88% in the test bacteria. Biofilm reduction was also examined using SEM and Confocal Laser Scanning Microscopy (CLSM). Interaction of IONPs with bacterial cells generated ROS contributing to reduced biofilm formation. The present study for the first time report that these IONPs were effective in obliterating pre-formed biofilms. Thus, it is envisaged that these nanoparticles with broad-spectrum biofilm inhibitory property could be exploited in the food industry as well as in medical settings to curtail biofilm based infections and losses.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Nunc® MicroWell 96 well polystyrene plates, polystyrene, clear, round bottom, 300 μL/well, sterile, 50/cs