Merck
New potent dual inhibitors of CK2 and Pim kinases: discovery and structural insights.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2010-04-20)
Miriam López-Ramos, Renaud Prudent, Virginie Moucadel, Céline F Sautel, Caroline Barette, Laurence Lafanechère, Liliane Mouawad, David Grierson, Frédéric Schmidt, Jean-Claude Florent, Panagis Filippakopoulos, Alex N Bullock, Stefan Knapp, Jean-Baptiste Reiser, Claude Cochet
ABSTRACT
Protein kinase casein kinase 2 (CK2) is a serine/threonine kinase with evidence of implication in growth dysregulation and apoptosis resistance, making it a relevant target for cancer therapy. Several CK2 inhibitors have been developed showing variable efficiency, emphasizing the need to expand the chemical diversity of those inhibitors. We report the identification and characterization of 2,8-difurandicarboxylic acid derivatives as a new class of nanomolar ATP-competitive inhibitors. Selectivity profiling pointed out proviral insertion Moloney virus kinases (Pim kinases) as the only other kinases that are significantly inhibited. By combining structure-activity relationship analysis with structural determination, we were able to determine the binding mode of these inhibitors for both kinases and to explain their strong inhibitory potency. Essential chemical features necessary for activity on both kinases were then identified. The described compounds are not cell permeable: however, they could provide a lead for developing novel inhibitors usable also in vivo. Given the similar but not redundant pathophysiological functions of CK2 and Pim family members, such inhibitors would provide new attractive leads for targeted cancer therapy. This work highlights that 2 functionally related kinases from different kinome branches display exquisite sensitivity to a common inhibitor.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Pim1 Protein, active, 10 µg, Active, N-terminal GST-tagged recombinant human full length Pim-1, for use in Kinase Assays.