Merck
PhenoFluorMix: practical chemoselective deoxyfluorination of phenols.
Organic letters (2015-01-27)
Teppei Fujimoto, Tobias Ritter
ABSTRACT
A practical deoxyfluorination with novel deoxyfluorinating reagent PhenoFluorMix, a mixture of N,N'-1,3-bis(2,6-diisopropylphenyl)chloroimidazolium chloride and CsF, is presented. PhenoFluorMix overcomes the challenges associated with hydrolysis of PhenoFluor. PhenoFluorMix does not hydrolyze, is readily available on decagram scale, and is storable in air. In this paper, we demonstrate the practicality of the reagent and exhibit the deoxyfluorination of a variety of phenols and heterocycles.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-Chloro-1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, ≥95%