Documenti
Z742395
PELCO® Sliding Latch Microslide Storage Boxes
yellow, Holds 100 x slides 76x26 mm
Sinonimo/i:
slide storage
About This Item
Codice UNSPSC:
41123302
NACRES:
MA.01
Materiali
acrylonitrile-butadiene-styrene (ABS)
yellow
Caratteristiche
Holds 100 x slides 76x26 mm
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Ted Pella 2302-5-CS
Lungh. × Largh. × Alt.
209 mm (8.23 in.) × 172 mm (6.77 in.) × 32 mm (1.26 in.)
Descrizione generale
Exceptional slide storage containers providing maximum protection for your microscope slides.
Caratteristiche e vantaggi
  • Numbered index sheet in lid for quick identification
  • Foam liner
  • Stackable
  • ABS-durable/impact resistant
  • Hinged lid
  • Unique sliding compression latch
Note legali
PELCO is a registered trademark of Ted Pella, Inc