Documenti
EP5702718009
Eppendorf® Adapter for 85/100 mL Round Bucket
pack of 2 ea
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.82
Caratteristiche
Holds 85 - 100 mL tubes
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Eppendorf® 5702718009
tecniche
centrifugation: suitable
Compatibilità
for use with 100 mL round buckets
Note legali
Eppendorf is a registered trademark of Eppendorf AG