Documenti fondamentali
Z511706
Sigma-Aldrich
AldraAmine trapping packets
volume 1,000-4,000 mL, large
Sinonimo/i:
AldraAmine Packets, AldraAmine Packs, Amine Trapping Packets
Scegli un formato
1 EA
CHF 31.50
5 EA
CHF 136.00
25 EA
CHF 494.00
CHF 31.50
Spedizione prevista il27 aprile 2025
Scegli un formato
Cambia visualizzazione
1 EA
CHF 31.50
5 EA
CHF 136.00
25 EA
CHF 494.00
About This Item
Codice UNSPSC:
24141516
NACRES:
NB.43
CHF 31.50
Spedizione prevista il27 aprile 2025
Capacità
1000-4000 mL
Volume
1,000-4,000 mL
Descrizione generale
Two common solvents used in peptide synthesis are dimethylformamide (DMF) and 1-methyl-2-pyrrolidinone (NMP). Both solvents will degrade naturally over time producing unwanted amine impurities that can contaminate your synthesis. AldraAmine trapping packets offer a solution to this problem by effectively reducing amine and water contaminants to levels near those of freshly distilled solvent. Just insert a packet into the bottle of solvent and let it do the rest. Packets are made of an ultra-high-molecular-weight polyethylene membrane that has been tested for safe long-term exposure to DMF and NMP. Packets are vacuum sealed.
  • Effectively reduces amine and water impurities for up to 3 months
  • Each packet individually vacuum sealed and fully activated for immediate use
  • Safe for automated synthesizers
  • Ideal for solid-phase and solution-phase peptide synthesis
Note: These traps are not designed to be used in aqueous solutions (greater than 500 ppm of water). Avoid exposure to solvents with a very high water content as heat may be produced.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)