Documenti
TEM-CF200-NI-TH
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
Thick, size 200 mesh, box of 50
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
box of 50
Misura
200 mesh
Descrizione generale
A 10 nm thick film of pure carbon supported standard squre mesh grids. Nickel grid, 200 mesh.