Documenti
SYNS111420
Synthware PTFE stopper with handle
joint: ST/NS 14/20
Sinonimo/i:
Synthware S111420, keyhead stopper
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.51
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware S111420
Descrizione generale
PTFE handle stoppers for easy gripping.
Note legali
Synthware is a trademark of Kemtech America Inc.