Documenti
SYNA882440
Synthware vacuum/inert gas adapter with T-bore glass stopcock
joint: ST/NS 24/40
Sinonimo/i:
Synthware A882440, T-shape vacuum gas adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A882440
Descrizione generale
With 4 mm T-bore glass stopcock having constant pressure retention device or 2 mm T-bore 1:5 taper PTFE plug.
Note legali
Synthware is a trademark of Kemtech America Inc.