Key Documents
BR6790
BRAND® valve set for seripettor®
Sinonimo/i:
Accessories, Bottle-top Dispenser, Dispenser, Valve set, seripettor®
About This Item
Codice UNSPSC:
41121806
NACRES:
NB.01
Confezionamento
pack of 1 set
Produttore/marchio commerciale
BRAND 6790
Descrizione generale
Includes - 1 filling valve body, 1 filling valve, 1 discharge valve, and 2 sealing rings
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG