- Up-regulation of FOS-like antigen 1 contributes to neuronal apoptosis in the cortex of rat following traumatic brain injury.
Up-regulation of FOS-like antigen 1 contributes to neuronal apoptosis in the cortex of rat following traumatic brain injury.
Metabolic brain disease (2017-10-29)
Xide Xu, Rui Jiang, Peipei Gong, Qianqian Liu, Yinan Chen, Shiqiang Hou, Debin Yuan, Jiansheng Shi, Qing Lan
PMID29080084
RÉSUMÉ
Neuronal apoptosis is an important process of secondary brain injury which is induced by neurochemical signaling cascades after traumatic brain injury (TBI). Present study was designed to investigate whether FOS-like antigen 1 (Fra-1) is involved in the neuronal apoptosis. Western blot analysis and immunohistochemistry in a rat TBI model revealed a significant increase in the expression of Fra-1 in the ipsilateral brain cortex, which was in parallel with increase in the expression of active caspase-3. With immunofluorescence double-labeling, Fra-1 was colocalized with active caspase-3 and with NeuN, a neuronal marker. In an in vitro cell injury model, H