Merck
Prostaglandin actions in established insect cell lines.
In vitro cellular & developmental biology. Animal (2017-04-30)
Yao-Fa Li, Hongwei Zhang, Joseph A Ringbauer, Cynthia L Goodman, Tamra Reall Lincoln, Kaile Zhou, David Stanley
RÉSUMÉ
Prostaglandins (PGs) are oxygenated metabolites of arachidonic acid (AA) and two other C20 polyunsaturated fatty acids that serve as biochemical signals mediating physiological functions. We reported that PGs influence protein expression in insect cell lines, which prompted the question: do PGs influence cell proliferation or viability in insect cell lines? Here, we report on the outcomes of experiments designed to address the question in cell lines from three insect orders: Hemiptera (squash bug, Anasa tristis, BCIRL-AtE-CLG15A), Coleoptera (red flour beetle, Tribolium castaneum, BCIRL-TcA-CLG1), and Lepidoptera (tobacco budworm, Heliothis virescens, BCIRL-HvAM1). Treating the insect cell lines with PGA
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %
Sigma-Aldrich
Methyl arachidonyl fluorophosphonate, ≥98%, methyl acetate solution (10 mg/mL)