Merck
Structural and Crystallographic Information from
Inorganic chemistry (2017-08-05)
Peter Werhun, David L Bryce
RÉSUMÉ
Despite the significance of nickel compounds, NMR spectroscopy of the active nickel isotope