Merck
Roles and tissue source of adiponectin involved in lifestyle modifications.
The journals of gerontology. Series A, Biological sciences and medical sciences (2012-05-09)
Yang Dai, Jing Pang, Huan Gong, Wei Fan, Tie-Mei Zhang
RÉSUMÉ
Calorie restriction and endurance exercise are known robust lifestyle modifications that delay the onset of type 2 diabetes and metabolic syndrome, however, its protective mechanism needs to be elucidated. To investigate the role of adiponectin in lifestyle modifications, male Sprague-Dawley rats were divided into groups of caloric restriction, exercise, and control for a 6-month intervention. Tissue and serum adiponectin levels, tissue expression of SIRT1, AMP-activated protein kinase (AMPK)α phosphorylation and AdipoR1, and insulin sensitivity were determined. All effects of adiponectin found in vivo were confirmed by L6 myoblast cells cultured with serum from a rat that received an intervention or by L6 cells with an AdipoR1 knockdown. Circulating adiponectin levels increased twofold to threefold in those rats in the caloric restriction and aerobic exercise groups, and adiponectin expression increased significantly not only in adipose tissue but also in skeletal muscle. The enhancement of SIRT1, AdipoR1 expression, and AMPKα phosphorylation in the skeletal muscle of the rats that underwent an intervention was simulated in the L6 myoblast cells cultured with serum from the intervention rats. The transferable effects of adiponectin in the serum were confirmed by blunting these effects in L6 myoblast cells upon knockdown of AdipoR1 or neutralizing the serum with an anti-adiponectin antibody. Adiponectin also exhibited a dose-dependent induction of its own receptor. The induction of AdipoR1 and SIRT1 expression and AMPKα phosphorylation by adiponectin was blunted when AMPKα, SIRT1, or AdipoR1, respectively, were knocked down. An elevated muscle-derived adiponectin can be attributed to lifestyle modifications. Adiponectin, which triggers the adiponectin receptor1 (AdipoR1) and its downstream targets AMPKα and SIRT1, was involved in the lifestyle modifications and control of type 2 diabetes.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Adiponectin (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution